4-(4-Methylpiperazin-1-yl)butanoic acid 95%

Supplier: Ambeed
Warning

A192650-1G A192650-250MG A192650-5G
76974-916EA 43 USD
76974-916 76974-918 77881-427
4-(4-Methylpiperazin-1-yl)butanoic acid 95%
4-(4-Methylpiperazin-1-yl)butanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD09717495
CAS Number: 58077-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR