2-Amino-7-bromo-1,3-benzothiazole 96%

Supplier: Ambeed
Warning

Synonyms: 7-Bromo-1,3-benzothiazol-2-amine, 7-Bromobenzo[d]thiazol-2-amine

A533805-250MG A533805-1G
76980-122EA 76 USD
76980-122 77673-428
2-Amino-7-bromo-1,3-benzothiazole 96%
2-Amino-7-bromo-1,3-benzothiazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂S
MDL Number: MFCD11110957
CAS Number: 20358-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR