2-(Methylsulfonyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminophenyl methyl sulphone

A928050-100MG A928050-1G A928050-250MG A928050-25G A928050-5G
76981-986EA 26.53 USD
76981-986 76981-988 76981-990 76981-992 76981-994
2-(Methylsulfonyl)aniline 97%
2-(Methylsulfonyl)aniline

Precaución: for Research Use Only

Formula: C₇H₉NO₂S
MW: 171.217 g/mol
MDL Number: MFCD08444390
CAS Number: 2987-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR