4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (4-(4-(tert-Butoxycarbonyl)piperazine-1-carbonyl)phenyl)boronic acid, 1-(1,1-Dimethylethyl) 4-(4-boronobenzoyl)-1-piperazinecarboxylate

A614036-1G A614036-10G
76980-566EA 30 USD
76980-566 77673-658
4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid 96%
4-(4-Boc-1-piperazinylcarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₃BN₂O₅
MDL Number: MFCD12025996
CAS Number: 1150114-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR