tert-Butyl (2-(isopropylamino)ethyl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A779239-100MG
A779239-1G
A779239-250MG
76981-572EA
32
USD
76981-572
76981-574
76981-576
tert-Butyl (2-(isopropylamino)ethyl)carbamate 98%
tert-Butyl (2-(isopropylamino)ethyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₂₂N₂O₂ |
MDL Number:
MFCD12828782 CAS Number: 320580-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...