(±)-2-Methylcaproic acid 98%

Supplier: Ambeed
Danger

A245009-25G A245009-5G
76976-560EA 59 USD
76976-560 76976-562
(±)-2-Methylcaproic acid 98%
(±)-2-Methylcaproic acid

Precaución: for Research Use Only

Formula: C₇H₁₄O₂
MDL Number: MFCD00002674
CAS Number: 4536-23-6
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR