5-Fluoroisophthalonitrile 98%
Supplier: Ambeed
|
Warning
|
A107443-100G
A107443-10G
A107443-1G
A107443-250MG
A107443-25G
A107443-5G
76970-490EA
451.95
USD
76970-490
76970-492
76970-494
76970-496
76970-498
76970-500
5-Fluoroisophthalonitrile 98%
5-Fluoroisophthalonitrile
Precaución: for Research Use Only
Formula:
C₈H₃FN₂ |
MDL Number:
MFCD05663831 CAS Number: 453565-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...