Ethyl 3-amino-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A115984-100MG A115984-1G A115984-250MG A115984-5G
76971-162EA 20 USD
76971-162 76971-164 76971-166 76971-168
Ethyl 3-amino-2-nitrobenzoate 98%
Ethyl 3-amino-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MDL Number: MFCD12911920
CAS Number: 193014-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR