Tivozanib 98+%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Chloro-4-((6,7-dimethoxyquinolin-4-yl)oxy)phenyl)-3-(5-methylisoxazol-3-yl)urea, AV-951

A770204-1MG A770204-5MG
76981-492EA 11.67 USD
76981-492 76981-494
Tivozanib 98+%
Tivozanib

Precaución: for Research Use Only

Formula: C₂₂H₁₉ClN₄O₅
MW: 454.9 g/mol
MDL Number: MFCD15146788
CAS Number: 475108-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR