5-(2-Methoxyethoxy)pyrazin-2-amine 95%

Supplier: Ambeed
Warning

A166294-100MG A166294-1G A166294-250MG A166294-5G
76973-808EA 98.67 USD
76973-808 76973-810 76973-812 76973-814
5-(2-Methoxyethoxy)pyrazin-2-amine 95%
5-(2-Methoxyethoxy)pyrazin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.1800 g/mol
MDL Number: MFCD13193283
CAS Number: 710322-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR