1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Acetylpyrazole-4-boronic Acid Pinacol Ester, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethanone, 1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]-ethanone

A787730-250MG A787730-5G A787730-1G
76981-584EA 35 USD
76981-584 76981-586 77882-695
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇BN₂O₃
MDL Number: MFCD09027064
CAS Number: 1150561-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR