1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate

A181194-10G A181194-1G A181194-250MG A181194-5G
76974-462EA 503.93 USD
76974-462 76974-464 76974-466 76974-468
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₁BN₂O₄
MW: 280.1278 g/mol
MDL Number: MFCD08706026
CAS Number: 864754-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR