4-Methyl-1H-pyrazolo[4,3-c]pyridine 95%

Supplier: Ambeed
Warning

A281722-100MG A281722-250MG A281722-1G
76977-422EA 101.85 USD
76977-422 76977-424 77672-452
4-Methyl-1H-pyrazolo[4,3-c]pyridine 95%
4-Methyl-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD11870061
CAS Number: 1140240-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR