N-Carbobenzoxy-DL-aspartic acid 98%

Supplier: Ambeed
Warning

Synonyms: Z-DL-Asp-OH, N-Cbz-DL-aspartic Acid, N-Benzyloxycarbonyl-DL-aspartic acid

A968048-100G A968048-25G A968048-5G
76982-032EA 62 USD
76982-032 76982-034 76982-036
N-Carbobenzoxy-DL-aspartic acid 98%
N-Carbobenzoxy-DL-aspartic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₆
MW: 267.24 g/mol
MDL Number: MFCD00063181
CAS Number: 4515-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR