2,3-Dibromobenzoic acid 98%

Supplier: Ambeed
Warning

A104286-1G A104286-250MG A104286-5G
76970-320EA 31 USD
76970-320 76970-322 77670-508
2,3-Dibromobenzoic acid 98%
2,3-Dibromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Br₂O₂
MDL Number: MFCD11520726
CAS Number: 603-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR