3-Chloro-2,6-difluorobenzonitrile 96%

Supplier: Ambeed
Danger

A998059-1G A998059-25G A998059-10G
76982-094EA 7 USD
76982-094 76982-096 77674-446
3-Chloro-2,6-difluorobenzonitrile 96%
3-Chloro-2,6-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₂N
MDL Number: MFCD06660172
CAS Number: 86225-73-2
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR