2,4-Dichloro-5-cyanothiazole 95%

Supplier: Ambeed
Warning

A296866-5G A296866-1G
76977-674EA 79 USD
76977-674 77881-818
2,4-Dichloro-5-cyanothiazole 95%
2,4-Dichloro-5-cyanothiazole

Precaución: for Research Use Only

Formula: C₄Cl₂N₂S
MDL Number: MFCD08669569
CAS Number: 82554-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR