Methyl-4-chloro-3-fluorobenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-chloro-3-fluorobenzoate, 4-Chloro-3-fluorobenzoic acid methyl ester
A280852-1G
A280852-25G
A280852-5G
76977-398EA
9
USD
76977-398
76977-400
76977-402
76977-404
Methyl-4-chloro-3-fluorobenzoate 98%
Methyl-4-chloro-3-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₆ClFO₂ |
MDL Number:
MFCD00209299 CAS Number: 206362-87-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...