6-Iodobenzo[d]thiazol-2-amine 95%

Supplier: Ambeed
Warning

A483336-100MG A483336-1G A483336-250MG
76979-754EA 46 USD
76979-754 76979-756 76979-758
6-Iodobenzo[d]thiazol-2-amine 95%
6-Iodobenzo[d]thiazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅IN₂S
MDL Number: MFCD00996639
CAS Number: 16582-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR