2,3-Dimethylpyridin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2,3-dimethylpyridine

A447697-100MG A447697-1G A447697-250MG A447697-5G
76979-460EA 20.16 USD
76979-460 76979-462 76979-464 76979-466
2,3-Dimethylpyridin-4-amine 97%
2,3-Dimethylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.168 g/mol
MDL Number: MFCD00030819
CAS Number: 122475-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR