6-Methylbenzo[d]isoxazol-3(2H)-one 98%

Supplier: Ambeed
Warning

A276555-100MG A276555-250MG
76977-294EA 19 USD
76977-294 76977-296
6-Methylbenzo[d]isoxazol-3(2H)-one 98%
6-Methylbenzo[d]isoxazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MDL Number: MFCD13180536
CAS Number: 66571-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR