5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrazolo[3,4-b]pyridine-5-boronic acid, pinacol ester, 1H-Pyrazolo[3,4-b]pyridine-5-boronic acid pinacol ester

A136781-100MG A136781-1G A136781-250MG A136781-5G
76972-366EA 42.44 USD
76972-366 76972-368 76972-370 77670-960
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine 95%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BN₃O₂
MW: 245.0853 g/mol
MDL Number: MFCD09832892
CAS Number: 1093819-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR