4-Chloro-5-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A113474-100MG A113474-1G A113474-250MG A113474-5G
76971-012EA 26.53 USD
76971-012 76971-014 76971-016 76971-018
4-Chloro-5-methoxypyrimidine 98%
4-Chloro-5-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.5600 g/mol
MDL Number: MFCD09966063
CAS Number: 695-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR