Methyl-2-fluoronicotinate 95%

Supplier: Ambeed
Warning

A234299-10G A234299-1G A234299-5G A234299-100G A234299-25G
76976-270EA 44 USD
76976-270 76976-272 76976-274 77672-088 77881-606
Methyl-2-fluoronicotinate 95%
Methyl-2-fluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD03095072
CAS Number: 446-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR