2-Methoxy-6-methylbenzonitrile 98%

Supplier: Ambeed
Warning

A380122-10G A380122-1G A380122-250MG A380122-5G
76978-828EA 177 USD
76978-828 76978-830 76978-832 76978-834
2-Methoxy-6-methylbenzonitrile 98%
2-Methoxy-6-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD10699539
CAS Number: 53005-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR