3-Methoxy-1H-indole 95%

Supplier: Ambeed
Warning

A345497-100MG A345497-1G A345497-250MG A345497-50MG
76978-454EA 170 USD
76978-454 76978-456 77881-949 77881-950
3-Methoxy-1H-indole 95%
3-Methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD12924274
CAS Number: 16712-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR