4-Methyloxazole 97%

Supplier: Ambeed
Danger

A140742-100MG A140742-1G A140742-250MG A140742-25G A140742-5G
76972-596EA 7.43 USD
76972-596 76972-598 76972-600 76972-602 76972-604
4-Methyloxazole 97%
4-Methyloxazole

Precaución: for Research Use Only

Formula: C₄H₅NO
MW: 83.089 g/mol
MDL Number: MFCD11110237
CAS Number: 693-93-6
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR