Chromane 98%

Supplier: Ambeed
Warning

Synonyms: Chroman

A403805-100MG A403805-1G A403805-5G A403805-250MG
76979-094EA 25 USD
76979-094 76979-096 76979-098 77882-073
Chromane 98%
Chromane

Precaución: for Research Use Only

Formula: C₉H₁₀O
MDL Number: MFCD00138123
CAS Number: 493-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR