3,4-Dimethylbenzyl bromide 97%

Supplier: Ambeed
Danger

A704709-10G A704709-1G A704709-250MG
76981-128EA 590 USD
76981-128 76981-130 77882-565
3,4-Dimethylbenzyl bromide 97%
3,4-Dimethylbenzyl bromide

Precaución: for Research Use Only

Formula: C9H11Br
MDL Number: MFCD06136695
CAS Number: 94416-66-7
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR