cis-3-Hydroxycyclobutanecarboxylic acid 98%

Supplier: Ambeed
Warning

A344168-1G A344168-250MG A344168-5G
76978-442EA 18 USD
76978-442 76978-444 77881-946
cis-3-Hydroxycyclobutanecarboxylic acid 98%
cis-3-Hydroxycyclobutanecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
MDL Number: MFCD12964956
CAS Number: 552849-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR