Favipiravir 99+%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide, T-705, 6-Fluoro-3-hydroxypyrazine-2-carboxamide

A265413-100G A265413-100MG A265413-1G A265413-250MG A265413-25G A265413-50MG A265413-5G
76977-074EA 360.71 USD
76977-074 76977-076 76977-078 76977-080 76977-082 76977-084 76977-086
Favipiravir 99+%
Favipiravir

Precaución: for Research Use Only

Formula: C₅H₄FN₃O₂
MW: 157.09 g/mol
MDL Number: MFCD16879084
CAS Number: 259793-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR