3-Aminooxetane-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A115749-10G A115749-250MG A115749-5G A115749-1G
76971-148EA 777 USD
76971-148 76971-150 76971-152 77566-088
3-Aminooxetane-3-carboxylic acid 95%
3-Aminooxetane-3-carboxylic acid

Precaución: For research use only.

Formula: C₄H₇NO₃
MDL Number: MFCD11035888
CAS Number: 138650-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR