Diethyl-3,4-pyridinedicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Cinchomeronic ester, diethyl pyridine-3,4-dicarboxylate, ethyl 4-(ethoxycarbonyl)pyridine-3-carboxylate

A729054-1G A729054-5G
76981-266EA 32.89 USD
76981-266 76981-268
Diethyl-3,4-pyridinedicarboxylate 98%
Diethyl-3,4-pyridinedicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₄
MW: 223.228 g/mol
MDL Number: MFCD00012349
CAS Number: 1678-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR