tert-Butyl 6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate 98%

Supplier: Ambeed
Warning

A236307-1G A236307-250MG A236307-100MG
76976-340EA 658.82 USD
76976-340 76976-342 77612-306
tert-Butyl 6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate 98%
tert-Butyl 6-amino-3-azabicyclo[3.1.0]hexane-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
MDL Number: MFCD09040607
CAS Number: 208837-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR