2-Chloro-8-nitroquinoline 95%

Supplier: Ambeed
Warning

A745081-1G A745081-250MG A745081-5G
76981-342EA 115.64 USD
76981-342 76981-344 76981-346
2-Chloro-8-nitroquinoline 95%
2-Chloro-8-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MW: 208.60 g/mol
MDL Number: MFCD00160589
CAS Number: 4225-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR