Plerixafor (AMD3100) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
AMD 3100, Mezobil, JM 3100, Plerixafor, 1,4-Bis((1,4,8,11-tetraazacyclotetradecan-1-yl)methyl)benzene, 1,1'-(1,4-phenylenebis(methylene))bis(1,4,8,11-tetraazacyclotetradecane)octahydrochloride dihydrate, 1,4-Bis[(1,4,8,11-tetraazacyclotetradecan-1-yl)methyl]benzene
A195007-100MG
A195007-10MG
A195007-1MG
A195007-250MG
76975-002EA
102.91
USD
76975-002
76975-004
76975-006
77611-872
Plerixafor (AMD3100) 99%
Plerixafor (AMD3100)
Precaución: for Research Use Only
Formula:
C₂₈H₅₄N₈ MW: 502.78 g/mol |
MDL Number:
MFCD05662218 CAS Number: 110078-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...