[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)-4'-methyldiphenyl ether hydrochloride, 4-(4-Methylphenoxy)benzylamine hydrochloride

A413552-100G A413552-25G A413552-5G
76979-184EA 84.88 USD
76979-184 76979-186 76979-188
[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride 98%
[4-(4-Methylphenoxy)phenyl]methylamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO·ClH
MW: 249.736 g/mol
MDL Number: MFCD02089411
CAS Number: 262862-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR