Methyl 3-bromo-5-methylbenzoate 97%

Supplier: Ambeed
Warning

A199774-100MG A199774-10G A199774-1G A199774-250MG A199774-25G
76975-198EA 5.31 USD
76975-198 76975-200 76975-202 76975-204 76975-206
Methyl 3-bromo-5-methylbenzoate 97%
Methyl 3-bromo-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
MDL Number: MFCD11007827
CAS Number: 478375-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR