1-(3-Chloropyrazin-2-yl)ethanone 98%
Supplier: Ambeed
|
Warning
|
A651802-100MG
A651802-1G
A651802-250MG
A651802-5G
76980-816EA
14
USD
76980-816
76980-818
76980-820
76980-822
1-(3-Chloropyrazin-2-yl)ethanone 98%
1-(3-Chloropyrazin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O |
MDL Number:
MFCD10697755 CAS Number: 121246-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...