Ethyl-1-boc-3-azetidineacetate 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Ethoxy-2-oxoethyl)azetidine-1-carboxylic acid tert-Butyl ester, Ethyl 1-(tert-Butoxycarbonyl)-3-azetidineacetate, 1-(tert-Butoxycarbonyl)-3-(2-ethoxy-2-oxoethyl)azetidine, 1-Boc-3-(2-ethoxy-2-oxoethyl)azetidine, tert-Butyl 3-(2-Ethoxy-2-oxoethyl)azetidine-1-carboxylate, Ethyl 1-Boc-3-azetidineacetate, 1-(tert-Butoxycarbonyl)-3-azetidineacetic acid ethyl ester

A257947-10G A257947-1G A257947-25G A257947-5G
76976-856EA 43 USD
76976-856 76976-858 76976-860 76976-862
Ethyl-1-boc-3-azetidineacetate 97%
Ethyl-1-boc-3-azetidineacetate

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD11616049
CAS Number: 158602-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR