2-Iodo-3-nitrobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A237196-1G
A237196-250MG
A237196-10G
76976-368EA
50.93
USD
76976-368
76976-370
77672-116
2-Iodo-3-nitrobenzoic acid 98%
2-Iodo-3-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄INO₄ MW: 293.02 g/mol |
MDL Number:
MFCD00277941 CAS Number: 5398-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...