3,6-Dihydro-2H-pyran 96%

Supplier: Ambeed
Danger

A758233-1G A758233-25G A758233-5G
76981-410EA 22.28 USD
76981-410 76981-412 76981-414
3,6-Dihydro-2H-pyran 96%
3,6-Dihydro-2H-pyran

Precaución: for Research Use Only

Formula: C₅H₈O
MW: 84.1164 g/mol
MDL Number: MFCD17171194
CAS Number: 3174-74-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR