2-Amino-3-chlorobenzylalkohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Amino-3-chlorophenyl)methanol

A184569-1G A184569-250MG A184569-5G
76974-560EA 65.78 USD
76974-560 76974-562 76974-564
2-Amino-3-chlorobenzylalkohol 98%
2-Amino-3-chlorobenzylalkohol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.6000 g/mol
MDL Number: MFCD13193432
CAS Number: 61487-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR