1,2,3,6-Tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

A765919-100MG A765919-1G
76981-454EA 33.95 USD
76981-454 76981-456
1,2,3,6-Tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
1,2,3,6-Tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₂₀BNO₂
MW: 209.09 g/mol
MDL Number: MFCD11506075
CAS Number: 375853-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR