1-Methyl-1,4-diazepane 95%

Supplier: Ambeed
Danger

Synonyms: 1-Methylhomopiperazine

A819097-100MG A819097-10G A819097-1G A819097-250MG A819097-25G A819097-5G
76981-730EA 5.31 USD
76981-730 76981-732 76981-734 76981-736 76981-738 76981-740
1-Methyl-1,4-diazepane 95%
1-Methyl-1,4-diazepane

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00059810
CAS Number: 4318-37-0
UN: 2920
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR