(Cyclopropylmethyl)(methyl)amine 95%

Supplier: Ambeed
Danger

A470085-100MG A470085-1G A470085-250MG A470085-5G
76979-650EA 49 USD
76979-650 76979-652 76979-654 76979-656
(Cyclopropylmethyl)(methyl)amine 95%
(Cyclopropylmethyl)(methyl)amine

Precaución: for Research Use Only

Formula: C₅H₁₁N
MDL Number: MFCD11150443
CAS Number: 18977-45-2
UN: 2733
ADR: 3,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR