4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,5-Dichloro-1,2,3-dithiazol-1-ium chloride
A649186-100MG
A649186-1G
A649186-250MG
A649186-5G
76980-802EA
11.67
USD
76980-802
76980-804
76980-806
76980-808
4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride 95%
4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride
Precaución: for Research Use Only
Formula:
C₂Cl₂NS₂·Cl MW: 208.517 g/mol |
MDL Number:
MFCD03426194 CAS Number: 75318-43-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...