GW 9508 98%

Supplier: Ambeed
Warning

Synonyms: GPR40 Agonist, GW9508, 4-[[(3-Phenoxyphenyl)methyl]amino]benzene propanoic acid, GW-9508, 3-(4-((3-Phenoxybenzyl)amino)phenyl)propanoic acid, 3-(4-(((3-(Phenoxy)phenyl)methyl)amino)phenyl)propanoic acid, Free Fatty Acid Receptor 1 Agonist I, FFA1 Agonist I

A144748-100MG A144748-10MG A144748-1MG A144748-25MG A144748-50MG A144748-5MG A144748-1G A144748-250MG
76972-764EA 234 USD
76972-764 76972-766 76972-768 76972-770 76972-772 76972-774 77881-126 77881-127
GW 9508 98%
GW 9508

Precaución: for Research Use Only

Formula: C₂₂H₂₁NO₃
MDL Number: MFCD09753282
CAS Number: 885101-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR