Dimethyl(N,N,N',N'-tetramethylethylenediamine)palladium(II) 98%

Supplier: Ambeed
Warning

A809851-100MG A809851-1G A809851-250MG
76981-688EA 68.96 USD
76981-688 76981-690 76981-692
Dimethyl(N,N,N',N'-tetramethylethylenediamine)palladium(II) 98%
Dimethyl(N,N,N',N'-tetramethylethylenediamine)palladium(II)

Precaución: for Research Use Only

Formula: C₈H₂₂N₂Pd
MW: 252.69 g/mol
MDL Number: MFCD11045096
CAS Number: 113822-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR