6-Chloro-3-methylpyrimidin-4(3H)-one 95%

Supplier: Ambeed
Warning

A290194-1G A290194-250MG A290194-25G A290194-5G
76977-558EA 76.39 USD
76977-558 76977-560 76977-562 76977-564
6-Chloro-3-methylpyrimidin-4(3H)-one 95%
6-Chloro-3-methylpyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.5600 g/mol
MDL Number: MFCD00955650
CAS Number: 101079-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR